Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080924.2 | NP_001074393.1 | 3 - 4 | 661 | 103 | |
NM_001290501.1 | NP_001277430.1 | 4 - 5 | 508 | 103 | ||
GenBank mRNA | AK133342.1 | - | 4 - 5 | 509 | 103 | |
AK163639.1 | - | 3 - 4 | 661 | 103 | ||
BC151080.1 | - | 4 - 5 | 382 | 9087993 | 103 | |
BC151083.1 | - | 4 - 5 | 382 | 9087996 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |