Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252250.1 | NP_001239179.1 | 5 - 6 | 1080 | 95 | |
NM_001252251.1 | NP_001239180.1 | 5 - 6 | 1080 | 95 | ||
NM_001252252.1 | NP_001239181.1 | 5 - 6 | 1080 | 95 | ||
NM_001252253.1 | NP_001239182.1 | 5 - 6 | 545 | 95 | ||
NM_010898.4 | NP_035028.2 | 5 - 6 | 1080 | 95 | ||
GenBank mRNA | AK017343.1 | - | 5 - 6 | 546 | 95 | |
AK045998.1 | - | 5 - 6 | 902 | 95 | ||
AK167685.1 | - | 2 - 3 | 171 | 95 | ||
BC005442.1 | - | 5 - 6 | 946 | 3496053 | 95 | |
DV654856.1 | - | 2 - 3 | 138 | 95 | ||
L22987.1 | - | 5 - 6 | 538 | 95 | ||
L22988.1 | - | 5 - 6 | 538 | 95 | ||
L22989.1 | - | 5 - 6 | 538 | 95 | ||
L27090.1 | - | 5 - 6 | 531 | 95 | ||
L27105.1 | - | 5 - 6 | 531 | 95 | ||
L28176.1 | - | 5 - 6 | 838 | 95 | ||
X74671.1 | - | 5 - 6 | 1097 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |