Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cobl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282993.1 | NP_001269922.1 | 3 - 4 | 689 | 123 | |
NM_001282994.1 | NP_001269923.1 | 3 - 4 | 520 | 123 | ||
NM_172496.3 | NP_766084.3 | 3 - 4 | 689 | 123 | ||
GenBank mRNA | AK049730.1 | - | 3 - 4 | 482 | 123 | |
AK122334.1 | - | 3 - 4 | 520 | 123 | ||
AK147366.1 | - | 3 - 4 | 690 | 123 | ||
AK147633.1 | - | 3 - 4 | 690 | 123 | ||
AY308745.1 | - | 3 - 4 | 529 | 123 | ||
BC145116.1 | - | 3 - 4 | 123 | |||
BC145118.1 | - | 3 - 4 | 9053666 | 123 | ||
BC150706.1 | - | 3 - 4 | 9087617 | 123 | ||
U26967.1 | - | 2 - 3 | 262 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |