Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010471.4 | NP_034601.1 | 1 - 2 | 78 | 77 | |
GenBank mRNA | AK002992.1 | - | 1 - 2 | 78 | 77 | |
BC049607.1 | - | 1 - 2 | 43 | 6595363 | 77 | |
BY129411.1 | - | 1 - 2 | 67 | 77 | ||
CX239660.1 | - | 2 - 3 | 173 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |