Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029219.1 | NP_083495.1 | 7 - 8 | 1594 | 111 | |
GenBank mRNA | AK015966.1 | - | 2 - 3 | 278 | 111 | |
AK183889.1 | - | 1 - 2 | 119 | 111 | ||
BC052529.1 | - | 1 - 2 | 196 | 111 | ||
BC117807.1 | - | 7 - 8 | 1152 | 111 | ||
BC139808.1 | - | 7 - 8 | 1155 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |