Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161822.1 | NP_001155294.1 | 4 - 5 | 474 | 80 | |
NM_019958.4 | NP_064342.1 | 3 - 4 | 404 | 80 | ||
GenBank mRNA | AF191555.1 | - | 3 - 4 | 346 | 80 | |
AK018279.1 | - | 3 - 4 | 388 | 80 | ||
AK030492.1 | - | 3 - 4 | 405 | 80 | ||
AK045770.1 | - | 4 - 5 | 441 | 80 | ||
AK046009.1 | - | 3 - 4 | 363 | 80 | ||
AK078745.1 | - | 3 - 4 | 363 | 80 | ||
AK163188.1 | - | 3 - 4 | 361 | 80 | ||
BC064782.1 | - | 3 - 4 | 332 | 30476405 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |