Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000013084.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013084.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013084.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013102.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013102.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013102.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013093.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013093.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013093.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013086.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013086.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013086.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013091.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013091.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013091.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013101.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013101.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013101.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013087.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013087.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013087.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013090.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013090.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013090.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013092.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013092.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013092.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013097.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013097.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013097.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013085.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013085.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013085.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013088.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013088.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013088.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013089.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013089.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013089.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013095.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013095.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013095.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013099.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013099.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013099.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013094.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013094.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013094.mm.1 |
TC1000001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000013100.mm.1 |
NM_001195036.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013100.mm.1 |
NM_001195036.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000013100.mm.1 |
TC1000001804.mm.1 |
MTA TC |