Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029100.2 | NP_083376.2 | 4 - 5 | 696 | 95 | |
GenBank mRNA | CX562316.1 | - | 3 - 4 | 373 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |