Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252642.1 | NP_001239571.1 | 1 - 2 | 231 | 73 | |
GenBank mRNA | AK038480.1 | - | 1 - 2 | 232 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Probe spans exons |