Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162533.1 | NP_001156005.1 | 7 - 8 | 651 | 81 | |
NM_025856.3 | NP_080132.1 | 3 - 4 | 315 | 81 | ||
GenBank mRNA | AK076962.1 | - | 3 - 4 | 280 | 81 | |
AK155132.1 | - | 7 - 8 | 572 | 81 | ||
BC019568.1 | - | 7 - 8 | 568 | 81 | ||
BC039777.1 | - | 3 - 4 | 505 | 81 | ||
BC053421.1 | - | 3 - 4 | 276 | 6706860 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |