Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145970.1 | NP_001139442.1 | 3 - 4 | 723 | 70 | |
NM_144941.3 | NP_659190.3 | 3 - 4 | 723 | 70 | ||
GenBank mRNA | AK151613.1 | - | 3 - 4 | 711 | 70 | |
BC023677.1 | - | 3 - 4 | 698 | 5354497 | 70 | |
BC023774.1 | - | 3 - 4 | 720 | 70 | ||
BQ713545.1 | - | 2 - 3 | 92 | 70 | ||
CV563343.1 | - | 2 - 3 | 294 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |