Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153402.2 | NP_700451.2 | 14 - 15 | 2098 | 81 | |
GenBank mRNA | AB081473.1 | - | 14 - 15 | 1950 | 81 | |
AK035042.1 | - | 14 - 15 | 2062 | 81 | ||
AK083652.1 | - | 14 - 15 | 2065 | 81 | ||
AK154436.1 | - | 14 - 15 | 2099 | 81 | ||
AK171118.1 | - | 14 - 15 | 2099 | 81 | ||
BC137964.1 | - | 14 - 15 | 1945 | 8860983 | 81 | |
BC145063.1 | - | 13 - 14 | 1808 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |