Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177077.2 | NP_796051.2 | 8 - 9 | 1023 | 85 | |
GenBank mRNA | AK090104.1 | - | 7 - 8 | 847 | 85 | |
AK144547.1 | - | 8 - 9 | 1024 | 85 | ||
BC144781.1 | - | 8 - 9 | 966 | 9053329 | 85 | |
BC145693.1 | - | 8 - 9 | 966 | 40130996 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |