Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600021091.mm.1 |
TC0600002714.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021091.mm.1 |
NM_173775.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021091.mm.1 |
NM_173775.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021099.mm.1 |
TC0600002714.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021099.mm.1 |
NM_173775.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021099.mm.1 |
NM_173775.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021105.mm.1 |
TC0600002714.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021105.mm.1 |
NM_173775.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021105.mm.1 |
NM_173775.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021092.mm.1 |
TC0600002714.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021092.mm.1 |
NM_173775.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021092.mm.1 |
NM_173775.3 |
Refseq |