Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zufsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028287.2 | NP_082563.1 | 3 - 4 | 759 | 72 | |
GenBank mRNA | AK046191.1 | - | 3 - 4 | 762 | 72 | |
AK052071.1 | - | 3 - 4 | 743 | 72 | ||
AK077730.1 | - | 3 - 4 | 780 | 72 | ||
AK160928.1 | - | 3 - 4 | 758 | 72 | ||
AK164346.1 | - | 1 - 2 | 2299 | 72 | ||
AK164710.1 | - | 1 - 2 | 2274 | 72 | ||
BC067065.1 | - | 2 - 3 | 673 | 72 | ||
BC089514.1 | - | 3 - 4 | 772 | 30252639 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |