Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001160.3 | NP_001001160.1 | 8 - 9 | 2514 | 61 | |
NM_001289674.1 | NP_001276603.1 | 7 - 8 | 2345 | 61 | ||
NM_001289675.1 | NP_001276604.1 | 7 - 8 | 2339 | 61 | ||
GenBank mRNA | AK129466.1 | - | 6 - 7 | 1891 | 61 | |
AK147230.1 | - | 8 - 9 | 2406 | 61 | ||
AK147681.1 | - | 7 - 8 | 2204 | 61 | ||
BC061475.1 | - | 5 - 6 | 986 | 61 | ||
BC070445.1 | - | 7 - 8 | 2345 | 30536738 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |