Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P66520 |
NM_177686.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P66520 |
AK084335.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P66520 |
NM_177686.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P66520 |
AK084335.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
142703 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
521562 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
579361 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
49655 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
885252 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
537462 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
640886 |
AB045293.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
640886 |
AK044384.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
640886 |
AK079893.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
640886 |
BC005722.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
640886 |
NM_021793.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
124714 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
87253 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P157242 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P122556 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P66520 |
NM_177686.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P66520 |
AK084335.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P66520 |
NM_177686.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P66520 |
AK084335.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842982 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842967 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842983 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842966 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842987 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842980 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842963 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842977 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842985 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842971 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842973 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842984 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842976 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842965 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842970 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24842968 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P122556 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06067735 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P157242 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P66520 |
NM_177686.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P66520 |
AK084335.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P66520 |
NM_177686.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P66520 |
AK084335.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P137354 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P157242 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P122556 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05775692 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05290089 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05469574 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842951 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843002 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843058 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843050 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842980 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842995 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842988 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842984 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24843103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24842968 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842995 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842957 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05697426 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842991 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842984 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843037 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842990 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843003 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843010 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842987 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842983 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842972 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842971 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843000 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842988 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842958 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843020 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05812082 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05053882 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842968 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842967 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842998 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842951 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842955 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842970 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842952 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843029 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843031 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843034 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842982 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843017 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842993 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05841790 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842950 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05182416 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843018 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843012 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842963 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843016 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05854213 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842953 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05290088 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842977 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843002 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842948 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24842980 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843011 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24843005 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05572932 |
- |
- |