Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600021955.mm.1 |
TC0600002843.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021955.mm.1 |
NM_001037539.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021955.mm.1 |
NM_001037539.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021955.mm.1 |
NM_001170419.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021955.mm.1 |
NM_001170419.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021955.mm.1 |
NM_015768.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021955.mm.1 |
NM_015768.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021949.mm.1 |
TC0600002843.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021949.mm.1 |
NM_001037539.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021949.mm.1 |
NM_001037539.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021949.mm.1 |
NM_001170419.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021949.mm.1 |
NM_001170419.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021949.mm.1 |
NM_015768.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021949.mm.1 |
NM_015768.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021954.mm.1 |
TC0600002843.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021954.mm.1 |
NM_001037539.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021954.mm.1 |
NM_001037539.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021954.mm.1 |
NM_001170419.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021954.mm.1 |
NM_001170419.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021954.mm.1 |
NM_015768.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021954.mm.1 |
NM_015768.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021950.mm.1 |
TC0600002843.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021950.mm.1 |
NM_001037539.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021950.mm.1 |
NM_001037539.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021950.mm.1 |
NM_001170419.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021950.mm.1 |
NM_001170419.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021950.mm.1 |
NM_015768.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021950.mm.1 |
NM_015768.2 |
Refseq |