Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabarapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020590.4 | NP_065615.1 | 3 - 4 | 548 | 219 | |
GenBank mRNA | AB041648.1 | - | 3 - 4 | 572 | 219 | |
AF180518.1 | - | 3 - 4 | 575 | 219 | ||
AK006103.1 | - | 3 - 4 | 347 | 219 | ||
AK011975.1 | - | 3 - 4 | 543 | 219 | ||
AK014083.1 | - | 3 - 4 | 362 | 219 | ||
AK015049.1 | - | 3 - 4 | 365 | 219 | ||
AK018687.1 | - | 3 - 4 | 549 | 219 | ||
AK159015.1 | - | 3 - 4 | 543 | 219 | ||
AK167500.1 | - | 3 - 4 | 541 | 219 | ||
AK168921.1 | - | 3 - 4 | 580 | 219 | ||
BC004602.1 | - | 3 - 4 | 483 | 3585132 | 219 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |