Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025995.2 | NP_080271.2 | 1 - 2 | 207 | 94 | |
GenBank mRNA | AK011820.1 | - | 1 - 2 | 205 | 94 | |
AK135916.1 | - | 1 - 2 | 162 | 94 | ||
AK160201.1 | - | 1 - 2 | 163 | 94 | ||
BC053434.1 | - | 1 - 2 | 131 | 6336178 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |