Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199306.4 | NP_955010.1 | 10 - 11 | 1362 | 64 | |
GenBank mRNA | AK131700.1 | - | 12 - 13 | 1518 | 64 | |
BC034833.1 | - | 3 - 4 | 285 | 5099225 | 64 | |
BC048824.1 | - | 10 - 11 | 1156 | 4188979 | 64 | |
BC057107.1 | - | 9 - 10 | 1005 | 5697877 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |