Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199306.4 | NP_955010.1 | 3 - 4 | 90 | ||
GenBank mRNA | AK131700.1 | - | 5 - 6 | 90 | ||
AK172431.1 | - | 3 - 4 | 552 | 90 | ||
BC057107.1 | - | 2 - 3 | 5697877 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |