Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tesk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146151.4 | NP_666263.3 | 8 - 9 | 1179 | 70 | |
GenBank mRNA | AK031814.1 | - | 8 - 9 | 1172 | 70 | |
AK047755.1 | - | 8 - 9 | 1180 | 70 | ||
AK053368.1 | - | 1244 | 70 | |||
AK134575.1 | - | 4 - 5 | 285 | 70 | ||
BC019149.1 | - | 7 - 8 | 912 | 5053345 | 70 | |
BC029766.1 | - | 8 - 9 | 1157 | 4947564 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |