Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145706.2 | NP_663752.2 | 4 - 5 | 565 | 65 | |
GenBank mRNA | AK011422.1 | - | 4 - 5 | 530 | 65 | |
AK090019.1 | - | 4 - 5 | 528 | 65 | ||
AK146262.1 | - | 4 - 5 | 567 | 65 | ||
BC052530.1 | - | 4 - 5 | 493 | 65 | ||
BF607996.1 | - | 3 - 4 | 333 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |