Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153055.3 | NP_694695.3 | 2 - 3 | 491 | 132 | |
GenBank mRNA | AK036629.1 | - | 2 - 3 | 186 | 132 | |
AK049737.1 | - | 2 - 3 | 182 | 132 | ||
AY238936.1 | - | 2 - 3 | 261 | 132 | ||
BC019366.1 | - | 2 - 3 | 255 | 5066908 | 132 | |
BC031846.1 | - | 2 - 3 | 255 | 5066822 | 132 | |
BC059816.1 | - | 2 - 3 | 491 | 132 | ||
BC080810.1 | - | 2 - 3 | 372 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |