Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177758.4 | NP_808426.2 | 4 - 5 | 854 | 78 | |
GenBank mRNA | AK047820.1 | - | 4 - 5 | 775 | 78 | |
AK153688.1 | - | 4 - 5 | 791 | 78 | ||
AK153845.1 | - | 4 - 5 | 791 | 78 | ||
BC076602.1 | - | 4 - 5 | 770 | 30611085 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |