Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011704.3 | NP_035834.2 | 5 - 6 | 1383 | 72 | |
GenBank mRNA | AJ132098.1 | - | 5 - 6 | 1216 | 72 | |
AK002773.1 | - | 5 - 6 | 1215 | 72 | ||
AK145984.1 | - | 5 - 6 | 1289 | 72 | ||
AK149485.1 | - | 3 - 4 | 764 | 72 | ||
AK167427.1 | - | 5 - 6 | 1215 | 72 | ||
BC019203.1 | - | 5 - 6 | 1209 | 5041670 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |