Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138628.3 | NP_619534.2 | 2 - 3 | 650 | 92 | |
GenBank mRNA | AF422244.1 | - | 2 - 3 | 587 | 92 | |
AF422245.1 | - | 2 - 3 | 587 | 92 | ||
AK052324.1 | - | 2 - 3 | 614 | 92 | ||
AK084579.1 | - | 2 - 3 | 650 | 92 | ||
AK084639.1 | - | 2 - 3 | 651 | 92 | ||
AK085904.1 | - | 2 - 3 | 544 | 92 | ||
AK137442.1 | - | 2 - 3 | 565 | 92 | ||
BC141023.1 | - | 2 - 3 | 563 | 9055635 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |