Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019549.2 | NP_062422.1 | 7 - 8 | 966 | 74 | |
GenBank mRNA | AF073294.1 | - | 7 - 8 | 950 | 74 | |
AF181829.1 | - | 7 - 8 | 910 | 74 | ||
AF303745.1 | - | 8 - 9 | 898 | 74 | ||
AK033872.1 | - | 7 - 8 | 904 | 74 | ||
AK038804.1 | - | 1004 | 74 | |||
AK050340.1 | - | 7 - 8 | 908 | 74 | ||
AK075810.1 | - | 7 - 8 | 910 | 74 | ||
AK138831.1 | - | 7 - 8 | 904 | 74 | ||
AK151460.1 | - | 7 - 8 | 878 | 74 | ||
AK152501.1 | - | 7 - 8 | 878 | 74 | ||
AK153109.1 | - | 7 - 8 | 878 | 74 | ||
AK156235.1 | - | 7 - 8 | 967 | 74 | ||
BC066185.1 | - | 7 - 8 | 892 | 30433827 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |