Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nprl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284359.1 | NP_001271288.1 | 4 - 5 | 528 | 64 | |
NM_181569.3 | NP_853547.1 | 3 - 4 | 489 | 64 | ||
NR_104306.1 | 3 - 4 | 407 | 64 | |||
GenBank mRNA | AK036405.1 | - | 3 - 4 | 398 | 64 | |
AK036633.1 | - | 2 - 3 | 458 | 64 | ||
AK047190.1 | - | 4 - 5 | 437 | 64 | ||
AK081902.1 | - | 3 - 4 | 400 | 64 | ||
AK154884.1 | - | 3 - 4 | 359 | 64 | ||
BC064471.1 | - | 3 - 4 | 489 | 6334386 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |