Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1994462 |
AK052712.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1994462 |
AJ310753.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1994462 |
AJ627045.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1994462 |
NM_019404.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P306933 |
AJ310753.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P306933 |
NM_019404.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P306933 |
AJ310753.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P306933 |
NM_019404.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
860289 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
AF183960.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
AF199491.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
AK041713.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
AK084516.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
AK087790.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
AK147356.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
AK147433.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
AK149340.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
NM_009834.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
U70139.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914725 |
XM_001477322.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
113958 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P142316 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P306933 |
AJ310753.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P306933 |
NM_019404.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P306933 |
AJ310753.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P306933 |
NM_019404.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1994462 |
AK052712.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1994462 |
AJ310753.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1994462 |
AJ627045.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1994462 |
NM_019404.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P306933 |
AJ310753.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P306933 |
NM_019404.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P306933 |
AJ310753.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P306933 |
NM_019404.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465321 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465323 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465310 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465284 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465327 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465294 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465318 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465301 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465317 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465289 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465311 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465306 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465277 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465296 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465286 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465298 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465285 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465300 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465324 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465280 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465290 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465292 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465303 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465326 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465315 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465313 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465320 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465282 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465278 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465325 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32465302 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P142316 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P306933 |
AJ310753.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P306933 |
NM_019404.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P306933 |
AJ310753.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P306933 |
NM_019404.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109179 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P142316 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14993944 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14993964 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14993909 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14993957 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14993941 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14993913 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14993946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465317 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465300 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465311 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465381 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465306 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465321 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465303 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465313 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465324 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465315 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465310 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465285 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465294 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465302 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465318 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465284 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465295 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465327 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465291 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32465380 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465318 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465327 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465317 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14993918 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465291 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465306 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465315 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465293 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465300 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14755338 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465313 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465311 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465298 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14993944 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14993912 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465290 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465288 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14993902 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14652137 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465320 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465324 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465303 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465321 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14993940 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465292 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465294 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32465325 |
- |
- |