Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00015043.mm.1 |
NM_001199351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015043.mm.1 |
NM_001199351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015043.mm.1 |
TC0X00002431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015046.mm.1 |
NM_001199351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015046.mm.1 |
NM_001199351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015046.mm.1 |
NM_012040.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015046.mm.1 |
NM_012040.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015046.mm.1 |
TC0X00002431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015041.mm.1 |
NM_001199351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015041.mm.1 |
NM_001199351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015041.mm.1 |
NM_001199352.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015041.mm.1 |
NM_001199352.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015041.mm.1 |
NM_012040.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015041.mm.1 |
NM_012040.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015041.mm.1 |
TC0X00002431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015045.mm.1 |
NM_001199351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015045.mm.1 |
NM_001199351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015045.mm.1 |
NM_012040.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015045.mm.1 |
NM_012040.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015045.mm.1 |
TC0X00002431.mm.1 |
MTA TC |