Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012060.4 | NP_036190.2 | 4 - 5 | 441 | 87 | |
GenBank mRNA | AK008043.1 | - | 4 - 5 | 441 | 87 | |
AK011500.1 | - | 4 - 5 | 442 | 87 | ||
AK151641.1 | - | 4 - 5 | 442 | 87 | ||
BC002106.1 | - | 4 - 5 | 409 | 3486784 | 87 | |
X81816.1 | - | 3 - 4 | 341 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |