Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmtk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005511.3 | NP_001005511.2 | 9 - 10 | 946 | 72 | |
NM_001290990.1 | NP_001277919.1 | 8 - 9 | 1101 | 72 | ||
GenBank mRNA | AB288873.1 | - | 9 - 10 | 940 | 72 | |
BC057032.1 | - | 8 - 9 | 834 | 72 | ||
BC083185.1 | - | 8 - 9 | 1101 | 6855048 | 72 | |
BC094377.1 | - | 9 - 10 | 946 | 30536822 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |