Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053196.3 | NP_444426.3 | 1 - 2 | 92 | 150 | |
GenBank mRNA | AK034875.1 | - | 92 | 150 | ||
AK149277.1 | - | 61 | 150 | |||
AK162407.1 | - | 1 - 2 | 92 | 150 | ||
BC019808.1 | - | 1 - 2 | 90 | 5043343 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |