Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps33a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029929.3 | NP_084205.3 | 2 - 3 | 206 | 106 | |
GenBank mRNA | AF439858.1 | - | 2 - 3 | 190 | 106 | |
AK019463.1 | - | 2 - 3 | 190 | 106 | ||
AK041982.1 | - | 2 - 3 | 202 | 106 | ||
AK140553.1 | - | 2 - 3 | 195 | 106 | ||
AK150282.1 | - | 2 - 3 | 198 | 106 | ||
AK150550.1 | - | 2 - 3 | 198 | 106 | ||
AK194964.1 | - | 2 - 3 | 120 | 106 | ||
BB852750.1 | - | 2 - 3 | 230 | 106 | ||
BC046417.1 | - | 2 - 3 | 197 | 4501207 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |