Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500012597.mm.1 |
TC0500001413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012597.mm.1 |
NM_029850.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012597.mm.1 |
NM_029850.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012590.mm.1 |
TC0500001413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012590.mm.1 |
NM_029850.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012590.mm.1 |
NM_029850.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012598.mm.1 |
TC0500001413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012599.mm.1 |
TC0500001413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012591.mm.1 |
TC0500001413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012591.mm.1 |
NM_029850.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012591.mm.1 |
NM_029850.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012592.mm.1 |
TC0500001413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012592.mm.1 |
NM_029850.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012592.mm.1 |
NM_029850.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012594.mm.1 |
TC0500001413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012594.mm.1 |
NM_029850.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012594.mm.1 |
NM_029850.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012596.mm.1 |
TC0500001413.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012596.mm.1 |
NM_029850.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012596.mm.1 |
NM_029850.3 |
Refseq |