Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013910.2 | NP_038938.1 | 5 - 6 | 921 | 77 | |
GenBank mRNA | AK031626.1 | - | 5 - 6 | 887 | 77 | |
AK129479.1 | - | 16 - 17 | 2358 | 77 | ||
BC057622.1 | - | 5 - 6 | 916 | 6402440 | 77 | |
BC065090.1 | - | 16 - 17 | 2557 | 6851421 | 77 | |
BC068239.1 | - | 13 - 14 | 2111 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |