Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klkb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008455.2 | NP_032481.2 | 8 - 9 | 967 | 81 | |
GenBank mRNA | BC026555.1 | - | 8 - 9 | 893 | 4972788 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |