Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145888.1 | NP_001139360.1 | 4 - 5 | 801 | 63 | |
NM_198644.2 | NP_941046.1 | 4 - 5 | 780 | 63 | ||
GenBank mRNA | BC053930.1 | - | 4 - 5 | 771 | 6389756 | 63 |
EU221277.1 | - | 4 - 5 | 631 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |