Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174850.3 | NP_777275.2 | 2 - 3 | 369 | 67 | |
GenBank mRNA | AB182579.1 | - | 2 - 3 | 220 | 67 | |
AK044547.1 | - | 2 - 3 | 220 | 67 | ||
AK085948.1 | - | 2 - 3 | 338 | 67 | ||
AK151086.1 | - | 2 - 3 | 368 | 67 | ||
AK164043.1 | - | 2 - 3 | 340 | 67 | ||
AK165558.1 | - | 2 - 3 | 219 | 67 | ||
AK196788.1 | - | 1 - 2 | 37 | 67 | ||
AK220308.1 | - | 2 - 3 | 146 | 67 | ||
BC043300.1 | - | 2 - 3 | 183 | 5025620 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |