Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026748.2 | NP_081024.3 | 33 - 34 | 5114 | 75 | |
GenBank mRNA | AB257854.1 | - | 33 - 34 | 5074 | 75 | |
AK148183.1 | - | 4 - 5 | 75 | |||
BC010333.1 | - | 4 - 5 | 695 | 75 | ||
BC063266.1 | - | 32 - 33 | 4882 | 6409930 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |