Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026748.2 | NP_081024.3 | 41 - 42 | 6262 | 68 | |
GenBank mRNA | AB257854.1 | - | 41 - 42 | 6222 | 68 | |
AK148183.1 | - | 12 - 13 | 1718 | 68 | ||
BC010333.1 | - | 12 - 13 | 1843 | 68 | ||
BC027811.1 | - | 1 - 2 | 786 | 68 | ||
BC051513.1 | - | 1 - 2 | 477 | 68 | ||
BC063266.1 | - | 40 - 41 | 6030 | 6409930 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |