Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033572.2 | NP_291050.1 | 9 - 10 | 1271 | 69 | |
GenBank mRNA | AF410456.1 | - | 9 - 10 | 1250 | 69 | |
AK005040.1 | - | 9 - 10 | 1218 | 69 | ||
AK017733.1 | - | 9 - 10 | 1270 | 69 | ||
BC024714.1 | - | 9 - 10 | 1243 | 5134834 | 69 | |
BC055335.1 | - | 9 - 10 | 1269 | 6412407 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |