Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199068.2 | NP_951031.2 | 4 - 5 | 1033 | 85 | |
GenBank mRNA | AK144418.1 | - | 4 - 5 | 926 | 85 | |
AK147375.1 | - | 4 - 5 | 1031 | 85 | ||
BC060238.1 | - | 4 - 5 | 1013 | 85 | ||
L26507.1 | - | 4 - 5 | 1009 | 85 | ||
U95016.1 | - | 4 - 5 | 1009 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |