Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199068.2 | NP_951031.2 | 7 - 8 | 1682 | 59 | |
GenBank mRNA | AK144418.1 | - | 7 - 8 | 1575 | 59 | |
AK147375.1 | - | 7 - 8 | 1680 | 59 | ||
BC002273.1 | - | 1 - 2 | 117 | 59 | ||
BC060238.1 | - | 7 - 8 | 1660 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |