Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmtk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081109.1 | NP_001074578.1 | 6 - 7 | 895 | 66 | |
GenBank mRNA | AB288872.1 | - | 6 - 7 | 723 | 66 | |
AK158724.1 | - | 6 - 7 | 896 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mus musculus Probe spans exons |