Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081066.1 | NP_001074535.1 | 17 - 18 | 3137 | 61 | |
GenBank mRNA | AK053114.1 | - | 7 - 8 | 1471 | 61 | |
AK173046.1 | - | 17 - 18 | 3037 | 61 | ||
BC132389.1 | - | 17 - 18 | 3025 | 40130666 | 61 | |
BC137791.1 | - | 17 - 18 | 3025 | 8860810 | 61 | |
BC144912.1 | - | 17 - 18 | 3025 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |