Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ermp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081213.1 | NP_001074682.1 | 11 - 12 | 2113 | 137 | |
GenBank mRNA | AK050284.1 | - | 11 - 12 | 2110 | 137 | |
AK081674.1 | - | 10 - 11 | 1630 | 137 | ||
AK129447.1 | - | 11 - 12 | 2041 | 137 | ||
AK137206.1 | - | 11 - 12 | 2069 | 137 | ||
BC031519.1 | - | 3 - 4 | 389 | 137 | ||
BC086627.1 | - | 11 - 12 | 2054 | 137 | ||
BC151181.1 | - | 11 - 12 | 2072 | 9088070 | 137 | |
BC151182.1 | - | 11 - 12 | 2072 | 9088071 | 137 | |
BC157914.1 | - | 11 - 12 | 2072 | 9088152 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |