Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900004630.mm.1 |
NM_001163635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004630.mm.1 |
NM_001163635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004630.mm.1 |
TC1900000521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004624.mm.1 |
NM_001163635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004624.mm.1 |
NM_001163635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004624.mm.1 |
TC1900000521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004623.mm.1 |
NM_001163635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004623.mm.1 |
NM_001163635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004623.mm.1 |
TC1900000521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004626.mm.1 |
NM_001163635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004626.mm.1 |
NM_001163635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004626.mm.1 |
TC1900000521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004631.mm.1 |
NM_001163635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004631.mm.1 |
NM_001163635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004631.mm.1 |
TC1900000521.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004625.mm.1 |
NM_001163635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004625.mm.1 |
NM_001163635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004625.mm.1 |
TC1900000521.mm.1 |
MTA TC |